accessibility menu, dialog, popup

UserName

NNHOOOOOHOOH(E/Z)

CAS RN 527680-57-5

IUPAC Name: N-{1-azabicyclo[2.2.2]octan-3-yl}-2,3-dihydro-1,4-benzodioxine-6-carboxamide; but-2-enedioic acid
Synonyms: N-{1-azabicyclo[2.2.2]octan-3-yl}-2,3-dihydro-1,4-benzodioxine-6-carboxamide; butenedioic acid
Molecular Weight (g/mol): 404.42
Molecular Formula: C20H24N2O7
InChi Key: QYQZUGYJVHHHEE-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.O=C(NC1CN2CCC1CC2)C1=CC2=C(OCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results