missing translation for 'onlineSavingsMsg'
Learn More

Promega pGEM™ Vectors DFS Item

Enable blue/white screening and contain both the SP6 and T7 RNA polymerase promoters

$204.23 - $258.25

Specifications

Promoter SP6, T7
Content And Storage -30°C to -10°C
Quantity 20 μg
For Use With (Application) Synthesis of RNA in-vitro and allows selection of recombinants by blue/white screening
View More Specs

Products 5
Catalog Number Mfr. No. Vector Price Quantity  
Catalog Number Mfr. No. Vector Price Quantity  
PR-P2271 DFS Item
View Documents
Promega
P2271
pGEM-3Zf(+) vector
Each for $204.23
Only null left
 
PR-P2261 DFS Item
View Documents
Promega
P2261
pGEM-3Zf(-) vector
Each for $204.23
Only null left
 
PR-P2241 DFS Item
View Documents
Promega
P2241
pGEM-5Zf(+) vector
Each for $258.25
Only null left
 
PR-P2251 DFS Item
View Documents
Promega
P2251
pGEM-7Zf(+) vector
Each for $258.25
Only null left
 
PR-P2151 DFS Item
View Documents
Promega
P2151
pGEM-3Z vector
Each for $204.23
Only null left
 
Description

Description

 

  • Multiple cloning site provides a selection of restriction sites and is flanked by the SP6 and T7 RNA polymerase promoters
  • lacZ α-peptide and multiple cloning site arrangement from pUC18 allow recombinants to be selected using blue/white screening
pGEM-3Z and -4Z Vectors†
  • Versatile vectors can be used for standard cloning and in vitro transcription
  • pGEM-3Z and pGEM-4Z vectors are identical except for the orientation of the SP6 and T7 promoters
pGEM-3Zf(+/-), -5Zf(+/-), -7Zf(+/-), -9Zf(-), -11Zf(+/-), and -13Zf(+) Vectors†
  • Vectors are derived from the pGEM-3Z vector and contain the origin of replication of the filamentous phage f1
  • Versatile vectors can be used for standard cloning, ssDNA production, and in vitro transcription
  • (+) and (-) vectors with the same numeric designation are identical except for the orientation of the f1 origin
  • pGEM-5Zf(+/-) and -7Zf(+/-) contain restriction site arrangements designed to generate unidirectional deletions with the Erase-a-Base™ System
  • pGEM-9Zf(-) permits the excision of an insert containing the SP6 and T7 polymerase promoters

 

Specifications

Specifications

SP6, T7
-30°C to -10°C
20 μg
Synthesis of RNA in-vitro and allows selection of recombinants by blue/white screening
Videos
SDS
Documents

Documents