Learn More
Promega pGEM™ Vectors
Enable blue/white screening and contain both the SP6 and T7 RNA polymerase promoters
Supplier: Promega P2261
Includes: Vector, 20μg; Bacterial Strain JM109, 500μL
Description
- Standard Cloning Vectors That Can Produce ssDNA and In vitro Transcribe Sense and Antisense RNA
- Blue/white screening for easy identification of recombinant clones
- In vitro transcription using the SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
- The difference between + and - vectors is orientation of the f1 origin
Specifications
| SP6, T7 | |
| -30°C to -10°C | |
| pGEM-3Zf(-) vector |
| EcoRI, SacI, KpnI, AvaI, SmaI, BamHI, XbaI, SalI, AccI, HincII, PstI, SphI and HindIII | |
| 20 μg | |
| Synthesis of RNA in-vitro and allows selection of recombinants by blue/white screening |