Learn More
Promega pGEM™ Vectors DFS Item
Enable blue/white screening and contain both the SP6 and T7 RNA polymerase promoters
Supplier:  Promega P2241
 Afficher plus de versions de ce produit
Numéro de catalogue. PR-P2241
Il en reste null
Découvrez plus d'offres spéciales
Comprend: Vector, 20μg; Bacterial Strain JM109, 500μL
Description
Description
  • Standard Cloning Vector and Template for In vitro Transcription
  • Blue/white screening to easily identify recombinant clones
  • Multiple cloning site provides a convenient selection of restriction sites for cloning
Spécifications
Spécifications
SP6, T7
-30°C to -10°C
pGEM-5Zf(+) vector
ApaI, AatII, SphI, NcoI, SacII, EcoRV, SpeI, NotI, PstI, SalI, NdeI, SacI, BstXI and NsiI
20 μg
Synthesis of RNA in-vitro and allows selection of recombinants by blue/white screening
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS