missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 04418948

Catalog No. 481810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
481810 10 mg
481850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 481810 Supplier Tocris Bioscience™ Supplier No. 4818/10

Potent and selective EP2 receptor antagonist

Potent EP2 receptor antagonist (IC50 = 16 nM for human EP2 receptors). Displays over 2000-fold selectivity for EP2 receptors over EP1, EP3, EP4, DP1 amd CRTH2 receptors; exhibits <30% binding at a diverse panel of GPCRs and ion channels at a concentration of 10 μM. Inhibits PGE2-induced increases in intracellular cAMP; reverses PGE2-invoked relaxation of mouse trachea (IC50 = 2.7 nM).

Chemical Identifiers

CAS 1078166-57-0
Molecular Formula C23H20FNO5

Specifications

Quantity 10 mg
Formula Weight 409.41
Purity 98%
Molecular Weight (g/mol) 409.413
Chemical Name or Material PF 04418948
Recommended Storage Store at -20°C