Learn More
Justrite PetroPad
Description
- Portable and easy to fold up and store without fear of leakage or mess
- Absorbs and permanently solidifies hydrocarbons while allowing rainwater to pass through
- Durable material and robust construction withstand the demands of a rigorous work environment
- Easily pressure wash built-up dirt and debris without releasing hydrocarbons
- TCLP leachate test and EPA Paint Filter Test (9095B) compliant
Spécifications
Spécifications
| Material | Polymer |
| Type | Sorbent, Oil-only |
| Disposable | Reusable |
| Weight Capacity | Heavyweight |
| Dimensions (L x W) | 53.9 x 35.8 in. |