missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, puriss., Honeywell™ Riedel-de Haën™

Puriss., low boiling point hydrogen treated naphtha, meets analytical specification of DAB, bpmin. 75% 40-60 °C (min. 75%)

Supplier:  Honeywell Chemicals 2458725L

 View more versions of this product

Catalog No. 60045647

Only null left
Explore available promotions

Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Specifications

Petroleum Ether
101316-46-5
2.5 L
Glass Bottle
UN1268
Meets analytical specification of DAB
0.642 to 0.656g/mL at 20°C
−40°C (−40°F)
MFCD00081849
puriss.

Chemical Identifiers

101316-46-5
MFCD00081849
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P273-P301 + P310-P331-P391-P403 + P235

H225-H304-H336-H411

DOTInformation : Class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S

EINECSNumber : 309-852-0

SDS
Documents

Documents

Promotions

Promotions