missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, Optima™ for HPLC and GC, Fisher Chemical™
Description
- Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
- Also meets ACS specifications
- Intended for research use only
Spécifications
Spécifications
| CAS | 8032-32-4 |
| Color | Colorless |
| Acidity | Pass Test |
| Quantity | 4 L |
| Identification | Pass Test |
| Packaging | Amber Soda Lime Glass |
| Physical Form | Liquid |
| Refractive Index | 1.3560 to 1.3660 (at 25°C) |
| Residue after Evaporation | 1ppm max. |
| UN Number | 1268 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. May cause heritable genetic damage. May cause eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air. NFPA Health:1 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |