missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, ACS, MilliporeSigma™

Catalog No. MPX04255
Change view
Click to view available options
Quantity:
1 L
4 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPX04255 4 L
MPX04252 1 L
MPX042520 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MPX04255 Supplier Technidata Supplier No. PX04255

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8032-32-4
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Color Colorless
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Petroleum Ether, For ACS analysis