missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum ether 35/60, HPLC Grade
Description
Petroleum ether 35/60 is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.640 g/mL |
| Flash Point | -49°C (-57°F) |
| Physical Form | Liquid |
| Refractive Index | 1.363 |
| Boiling Point | 35°C to 60°C |
| Grade | HPLC |
| Chemical Name or Material | Petroleum ether 35/60, HPLC Grade |
Safety and Handling
| Hazard Category | H225-H304-H315-H319-H336 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: II |
| EINECSNumber | 232-453-7 |
| RTECSNumber | OI6180000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only