missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Cleaner or Gum Solvent, 50/50 Toluene and Acetone, Fisher Chemical™

Catalog No. ST954
Catalog No. ST954 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. ST954

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An equal-volume blend of acetone and toluene to clean residue from test vessels and glassware prior to or after testing fuels, gasoline, and other petroleum-based samples. This blend is also known as gum solvent.

  • A 50/50 blend (by volume) of toluene and acetone
  • Suitable for use as a cleaner of residue after testing fuel and petroleum samples
  • This useful cleaner is pre-blended for added convenience and safety
  • Also known as gum solvent as referenced in ASTM D525
  • Packaged in 4L amber glass bottles

Specifications

CAS 67-64-1, 108-88-3
Color Clear, colorless
CAS Min % 49%
CAS Max % 51%
Quantity 4 L
Packaging Amber Glass Bottle
Specific Gravity 0.825 - 0.835 at 25°C
Grade Laboratory