missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Uniconazole, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 83657-22-1 |
| Physical Form | Neat |
| Linear Formula | C15H18 ClN3 O |
| MDL Number | MFCD01678674 |
| Synonym | (E)-(RS)-1-(4-Chlorophenyl)-4,4-dimethyl-2-(1 H-1,2,4-triazol-1-yl)pent-1-en-3-ol |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 291.78 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH