missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ TDCPP, MilliporeSigma™ Supelco™

Numéro de catalogue. 111018519
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111018519 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111018519 Fournisseur Merck Emd Millipore Code fournisseur 32951100MG
Il en reste null

TDCPP is classified under the chlorinated trialkyl organophosphate flame retardant (OPFR) group of compounds primarily utilized as additives in commercial products such as plastics, paints, textiles, furniture, etc.

Agriculture

Spécifications

Product Line PESTANAL
CAS 13674-87-8
Density 1.514 g/mL (at 20°C)
Flash Point 250°C to 252°C - open°Cup
Physical Form Neat
Linear Formula C9H15 Cl6O4 P
MDL Number MFCD00083121
Synonym Tris(1,3-dichloro-2-propyl) phosphate; 1,3-Dichloro-2-propanol phosphate; Phosphoric acid tris(1,3-dichloro-2-propyl ester)
Type Analytical Standard
Quantity 100 mg
Formula Weight 430.90
Grade Analytical Standard
Afficher plus de résultats Afficher moins de résultats

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH