missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Silthiofam, MilliporeSigma™ Supelco™

Catalog No. 111018341
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018341 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018341 Supplier Merck Emd Millipore Supplier No. 3249825MG

Silthiofam is a thiophenecarboxamide fungicide.

Agriculture

Specifications

Product Line PESTANAL
CAS 175217-20-6
Physical Form Neat
Linear Formula C13H21 NOSSi
MDL Number MFCD04112621
Synonym N-Allyl-4,5-dimethyl-2-(trimethylsilyl)thiophene-3-carboxamide
Type Analytical Standard
Quantity 25 mg
Formula Weight 267.46
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH