missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Pyrifenox, MilliporeSigma™ Supelco™

Catalog No. 111015746
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015746 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015746 Supplier Merck Emd Millipore Supplier No. 45737100MG

Pyrifenox is primarily used in controlling powdery mildews of field-grown fruits, mainly wine grapes, mango and nectarine trees.

Agriculture

Specifications

Product Line PESTANAL
CAS 88283-41-4
Flash Point 145°C
Physical Form Neat
Linear Formula C14H12 Cl2N2 O
MDL Number MFCD16661268
Type Analytical Standard
Quantity 100 mg
Formula Weight 295.16
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH