missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Propyzamide, MilliporeSigma™ Supelco™
Description
Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 23950-58-5 |
| Physical Form | Neat |
| Linear Formula | C12H11 Cl2 NO |
| MDL Number | MFCD00055346 |
| Synonym | Pronamide |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 256.13 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH