missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Propamocarb-(propyl-d7), ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111002499
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002499 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002499 Supplier Merck Emd Millipore Supplier No. 807575MG
Agriculture

Specifications

Product Line PESTANAL
CAS 1398065-89-8
Linear Formula C9D7H13N2O2
MDL Number MFCD04972577
Synonym Propamocarb-d7
Type Analytical Standard
Quantity 5 mg
Formula Weight 195.31
Percent Purity ≥98.0% (GC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH