missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Propachlor, MilliporeSigma™ Supelco™

Catalog No. 111019827
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019827 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019827 Supplier Merck Emd Millipore Supplier No. 45637250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Propachlor is a herbicide, widely used in oil seeds and crop planting.

Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 1918-16-7
Flash Point 100°C
Physical Form Neat
Linear Formula C11H14 ClNO
MDL Number MFCD00078731
Synonym 2-Chloro-N-isopropylacetanilide
Type Analytical Standard
Quantity 250 mg
Formula Weight 211.69
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH