missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Prochloraz Metabolite BTS44595, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 139520-94-8 |
| Linear Formula | C12H15 Cl3N2O2 |
| MDL Number | MFCD04125365 |
| Synonym | 1-Propyl-1-[2-(2,4,6-trichlorophenoxy)ethyl]urea |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 325.62 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH