missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Picric acid Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111018673
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018673 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018673 Supplier Merck Emd Millipore Supplier No. 4620910ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 88-89-1
Quantity 10 mL
Flash Point 2°C
Linear Formula (O2 N)3C6H2 OH
MDL Number MFCD00007102
Synonym 2,4,6-Trinitrophenol
Formula Weight 229.1
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH