missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Methfuroxam, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 28730-17-8 |
| Physical Form | Neat |
| Linear Formula | C14H15 NO2 |
| MDL Number | MFCD00055348 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 229.27 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH