missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Mesosulfuron-methyl, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 208465-21-8 |
| Physical Form | Neat |
| Linear Formula | C17H21N5O9S2 |
| MDL Number | MFCD04112616 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 503.51 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH