missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Mefluidide, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 53780-34-0 |
| Product Line | PESTANAL |
| Physical Form | Neat |
| Linear Formula | C11H13F3N2O3 S |
| MDL Number | MFCD01632752 |
| Synonym | 5′-(1,1,1-Trifluoromethanesulfonamido)acet-2′,4′-xylidide; 5-Acetamido-2,4-dimethyltrifluoromethanesulfonanilide; N-[2,4-Dimethyl-5-[[(trifluoromethyl)sulfonyl]amino]phenyl]acetamide |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 310.29 |
| Percent Purity | ≥98% (HPLC) |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH