missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Malathion Solution, 100 μg/mL in cycloHexane, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 121-75-5 |
| Flash Point | -18°C - Closed Cup |
| Linear Formula | C10H19O6 PS2 |
| MDL Number | MFCD00036233 |
| Type | Analytical Standard |
| Quantity | 2 mL |
| Formula Weight | 330.36 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 100 μg/mL in cyclohexane |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH