missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Isopyrazam, Mixture of isomers, MilliporeSigma™ Supelco™

Catalog No. 111018672
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018672 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018672 Supplier Merck Emd Millipore Supplier No. 32532100MG
Only null left

Isopyrazams are categorized under the class of succinate-dehydrogenase-inhibitor (SDHI) fungicides that are applied on a variety of fruits and vegetables.

Agriculture

Specifications

Product Line PESTANAL
CAS 881685-58-1
Physical Form Neat
Linear Formula C20H23F2N3 O
Synonym 3-(Difluoromethyl)-1-methyl-N-[1,2,3,4-tetrahydro-9-(1-methylethyl)-1,4-methanonaphthalen-5-yl]-1 H-pyrazole-4-carboxamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 359.41
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH