missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Iprovalicarb, Mixture of isomers, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 140923-17-7 |
| Physical Form | Neat |
| Linear Formula | C18H28N2O3 |
| MDL Number | MFCD04112614 |
| Synonym | Nalpha-(Isopropoxycarbonyl)-N-[1-(p-tolyl)ethyl]-L-valinamide; Isopropyl (S)-2-methyl-1-[1-(p-tolyl)ethylcarbamoyl]propylcarbamate |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 320.43 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH