missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Halofenozide, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018284
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018284 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018284 Supplier Merck Emd Millipore Supplier No. 6853525MG

Halofenozide is a systemic type of insect growth regulator belonging to the class of non-steroidal ecdysone agonists. It shows a broad spectrum of activity in controlling beetle grub and lepidopteran larval pests.

Specifications

CAS 112226-61-6
Product Line PESTANAL
Physical Form Neat
Linear Formula C18H19 ClN2O2
MDL Number MFCD03158909
Synonym N-tert-Butyl-N′-(4-chlorobenzoyl)benzohydrazide
Type Analytical Standard
Quantity 25 mg
Formula Weight 330.81
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) nitrogen content, 8.47%
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH