missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Fluxapyroxad, MilliporeSigma™ Supelco™

Catalog No. 111018109
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018109 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018109 Supplier Merck Emd Millipore Supplier No. 37047100MG

Fluxapyroxad belongs to carboxamides group of chemicals. It is a succinate dehydrogenase inhibitor (SDHI) in complex II of the mitochondrial respiratory chain.

Agriculture

Specifications

Product Line PESTANAL
CAS 907204-31-3
Physical Form Neat
Linear Formula C18H12F5N3 O
MDL Number MFCD22373628
Synonym 3-(Difluoromethyl)-1-methyl-N-(3 ′,4 ′,5 ′-trifluorobiphenyl-2-yl)pyrazole-4-carboxamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 381.30
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH