missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Fluazifop-P-butyl, MilliporeSigma™ Supelco™
Description
Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 79241-46-6 |
| Flash Point | 50°C - Closed Cup |
| Physical Form | Neat |
| Linear Formula | C19H20F3 NO4 |
| MDL Number | MFCD06199153 |
| Synonym | Butyl (R)-2-[4-(5-trifluoromethyl-2-pyridyloxy)phenoxy]propionate |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 383.36 |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH