missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Fenthion, MilliporeSigma™ Supelco™
Description
Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 55-38-9 |
| Physical Form | Neat |
| Linear Formula | C10H15O3 PS2 |
| MDL Number | MFCD00055449 |
| Synonym | O,O′-Dimethyl O′′-[3-methyl-4-(methylthio)phenyl] thiophosphate |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 278.33 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH