missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Fenoxaprop-P, MilliporeSigma™ Supelco™

Numéro de catalogue. 111012631
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111012631 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111012631 Fournisseur Merck Emd Millipore Code fournisseur 36850100MG
Il en reste null
Agriculture

Spécifications

Product Line PESTANAL
CAS 113158-40-0
Physical Form Neat
Linear Formula C16H12 ClNO5
MDL Number MFCD01311798
Synonym (R)-2-{4-[(6-Chlorobenzoxazol-2-yl)oxy]phenoxy}propionic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 333.72
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH