missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Famoxadone Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Numéro de catalogue. 111019540
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111019540 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111019540 Fournisseur Merck Emd Millipore Code fournisseur 334952MLR
Il en reste null

Famoxadone belongs to the oxazolidinedione class of fungicides used to control broad spectrum of fungal diseases in agricultural crops.

Agriculture

Spécifications

Product Line PESTANAL
CAS 131807-57-3
Flash Point 2°C
Linear Formula C22H18N2O4
MDL Number MFCD03427409
Synonym 3-Anilino-5-methyl-5-(4-phenoxyphenyl)oxazolidine-2,4-dione
Type Analytical Standard
Quantity 2 mL
Formula Weight 374.39
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile
Afficher plus de résultats Afficher moins de résultats

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH