missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Endrin aldehyde, MilliporeSigma™ Supelco™

Catalog No. 111015399
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015399 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015399 Supplier Merck Emd Millipore Supplier No. 442578

missing translation for 'validMainframeMsg'

Endrin aldehyde is an organochlorine type of pesticide.

Agriculture

Specifications

Packaging Ampule
Product Line PESTANAL
CAS 7421-93-4
Physical Form Neat
Type Analytical Standard
Quantity 25 mg
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH