missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ β-Endosulfan, MilliporeSigma™ Supelco™
Description
Agriculture and Environmental
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 33213-65-9 |
| Physical Form | Neat |
| Linear Formula | C9H6 Cl6O3 S |
| MDL Number | MFCD00151176 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 406.93 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH