missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Diethyl phthalate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019115
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019115 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019115 Supplier Merck Emd Millipore Supplier No. 530085MLF
Only null left

Diethyl phthalate belongs to the class of phthalates, widely used as solvents and odorless diluents in perfumes and cosmetics.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
Ignition Point 854°F
CAS 84-66-2
Vapor Pressure 1 mmHg (100°C)
Vapor Density 7.66 (vs. air)
Density 1.12 g/mL (at 25°C)
Flash Point 156°C
Physical Form Neat
Linear Formula C6H4-1,2-(CO2C2H5)2
MDL Number MFCD00009111
Melting Point -3°C
Boiling Point 298°C to 299°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 222.24
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH