missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Dichlormid, MilliporeSigma™ Supelco™

Catalog No. 111014089
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014089 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014089 Supplier Merck Emd Millipore Supplier No. 33613100MG

Dichlormid belongs to the class of dichloroacetamide safeners used to provide good protection on maize, wheat and rice grain against injury caused by pre-emergence applications of butachlor, metolachlor, chloroacetanilide and thiocarbamate.

Agriculture

Specifications

Product Line PESTANAL
CAS 37764-25-3
Physical Form Neat
Linear Formula C8H11 Cl2 NO
MDL Number MFCD00072464
Synonym N,N-Diallyldichloroacetamide; Dichloromid
Type Analytical Standard
Quantity 100 mg
Formula Weight 208.09
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH