missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Dibutyl phthalate-3,4,5,6-d4, MilliporeSigma™ Supelco™

Catalog No. 111018270
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018270 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018270 Supplier Merck Emd Millipore Supplier No. 3416925MG

Dibutyl phthalate-3,4,5,6-d4 may be used as a standard solution in the determination of phthalic acid esters in soil, using gas chromatography coupled with mass spectrometry (GC/MS).

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 93952-11-5
Physical Form Neat
Linear Formula C6D4-1,2-[CO2(CH2)3 CH3]2
MDL Number MFCD00144148
Melting Point -35°C
Boiling Point 340°C
Synonym Di-n-butyl phthalate-d4
Type Analytical Standard
Quantity 25 mg
Formula Weight 282.37
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH