missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ λ-Cyhalothrin, MilliporeSigma™ Supelco™

Catalog No. 111018491
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018491 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018491 Supplier Merck Emd Millipore Supplier No. 31058100MG

λ-Cyhalothrin is a type II highly active synthetic pyrethroid insecticide which contains a-cyano group.

Specifications

Product Line PESTANAL
CAS 91465-08-6
Physical Form Neat
Linear Formula C23H19 ClF3 NO3
MDL Number MFCD07370148
Type Analytical Standard
Quantity 100 mg
Formula Weight 449.85
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH