missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Benalaxyl-M, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 98243-83-5 |
| Physical Form | Neat |
| Linear Formula | C20H23 NO3 |
| MDL Number | MFCD11616749 |
| Synonym | N-(2,6-Dimethylphenyl)-N-(phenylacetyl)-D-alanine methyl ester |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 325.40 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH