missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Atrazine-desisopropyl, MilliporeSigma™ Supelco™

Catalog No. 111012280
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012280 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012280 Supplier Merck Emd Millipore Supplier No. 36628250MG

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Agriculture

Specifications

Product Line PESTANAL
CAS 1007-28-9
Physical Form Neat
Linear Formula C5H8 ClN5
MDL Number MFCD00078645
Synonym 2-Amino-4-chloro-6-ethylamino-1,3,5-triazine
Type Analytical Standard
Quantity 250 mg
Formula Weight 173.60
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH