missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Ametoctradin, MilliporeSigma™ Supelco™

Catalog No. 111015704
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015704 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015704 Supplier Merck Emd Millipore Supplier No. 3246150MG
Only null left

Ametoctradin is a novel triazolopyrimidine fungicide, considered as effective agricultural fungicide. It is also an oomycete-specific fungicide, as it is potent inhibitor of mitochondrial bc1 complex of oomycetes in mitochondrial respiration.

Agriculture

Specifications

Product Line PESTANAL
CAS 865318-97-4
Physical Form Neat
Linear Formula C15H25N5
Synonym 5-Ethyl-6-octyl[1,2,4 ]triazolo[1,5-a]pyrimidin-7-amine
Type Analytical Standard
Quantity 50 mg
Formula Weight 275.39
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH