missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Aldicarb-sulfone, MilliporeSigma™ Supelco™

Numéro de catalogue. 111018748
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111018748 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111018748 Fournisseur Merck Emd Millipore Code fournisseur 33387100MG
Il en reste null

Aldicarb-sulfone is a metabolite of the insecticide aldicarb used to control insects, mites and nematodes in agricultural crops.

Agriculture

Spécifications

Product Line PESTANAL
CAS 1646-88-4
Physical Form Neat
Linear Formula C7H14N2O4 S
MDL Number MFCD00041804
Melting Point 132°C to 135°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 222.26
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH