missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Acetochlor ESA sodium salt, MilliporeSigma™ Supelco™
Description
Agriculture and Environmental
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 947601-84-5 |
| Physical Form | Neat |
| Linear Formula | C14H20 NNaO5 S |
| MDL Number | MFCD09037339 |
| Synonym | 2-[(Ethoxymethyl)(2-ethyl-6-methylphenyl) amino]-2-oxo-ethanesulfonic acid sodium salt; Acetochlor ethanesulfonic acid sodium salt |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 337.37 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH