missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Acetaldehyde, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111020494
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020494 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020494 Supplier Merck Emd Millipore Supplier No. 506788

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetaldehyde is a highly inflammable liquid, which can be commercially produced from liquid-phase oxidation of ethylene. It can also be obtained as a co-product from the oxidation of saturated hydrocarbons like propane or butane. It can find applications as a chemical intermediate in the production of acetic acid.

Specifications

Packaging Ampule
CAS 75-07-0
Product Line PESTANAL
Vapor Pressure 14.63 psi (20°C)
Vapor Density 1.52 (vs. air)
Density 0.785 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3CHO
MDL Number MFCD00006991
Melting Point -125°C
Boiling Point 21°C
Synonym Ethanal
Type Food and Cosmetic Component Standard
Quantity 1000 mg
Formula Weight 44.05
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH