missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 3,5-Dichloroaniline Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022472
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022472 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022472 Supplier Merck Emd Millipore Supplier No. 36704100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 626-43-7
Melting Point 46°C to 52°C (literature)
Boiling Point 259°C to 260°C (741 mmHg) (literature)
Flash Point 133°C (Closed Cup)
Molecular Formula C6H5Cl2N
Linear Formula Cl2C6H3NH2
Quantity 100 mg
MDL Number MFCD00007774
UN Number UN 3442 6.1 / PGII
Formula Weight 162.02
Grade Analytical
Physical Form Neat
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH