missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ 2,5-Dichloroaniline Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | 2,5-Dichloroaniline |
| Melting Point | 47°C to 50°C (literature) |
| CAS | 95-82-9 |
| Quantity | 100 mg |
| Flash Point | 139°C |
| Packaging | Glass Ampule |
| Physical Form | Neat |
| Molecular Formula | C6H5Cl2N |
| Linear Formula | Cl2C6H3NH2 |
| MDL Number | MFCD00007667 |
| Show More |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH