missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 2,4-DDE Solution, 100 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020031
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020031 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020031 Supplier Merck Emd Millipore Supplier No. 366632ML
Agriculture

Specifications

Product Line PESTANAL
CAS 3424-82-6
Flash Point 11°C
Linear Formula C14H8 Cl4
MDL Number MFCD00036122
Synonym 2-(2-Chlorophenyl)-2-(4-chlorophenyl)-1,1-dichloroethene solution
Type Analytical Standard
Quantity 2 mL
Formula Weight 318.03
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in methanol
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH