missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ 2,3,4-Trichlorophenol, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 15950-66-0 |
| Physical Form | Neat |
| Linear Formula | Cl3C6H2 OH |
| MDL Number | MFCD00002161 |
| Melting Point | 75°C to 79°C; 75°C to 79°C |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 197.45 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH