missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 1,2-Dichlorobenzene Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022473
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022473 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022473 Supplier Merck Emd Millipore Supplier No. 36707100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 95-50-1
Melting Point -18°C to -17°C (literature)
Density 1.306 g/mL (at 25°C (literature))
Boiling Point 178°C to 180°C (literature)
Flash Point 66°C (Closed Cup)
Molecular Formula C6H4Cl2
Refractive Index n20/D 1.551 (literature)
Linear Formula C6H4Cl2
Quantity 100 mg
MDL Number MFCD00000535
UN Number UN 1591 6.1 / PGIII
Vapor Density 5.1 (vs air)
Vapor Pressure 1.6 mmHg (35°C)
Formula Weight 147.00
Grade Analytical Standard
Physical Form Neat
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH