missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Personal Protection (PPE) Labels - Cold Temperature Vinyl

Catalog No. 19111821
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19111821 10 x 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19111821 Supplier Brady™ Supplier No. 58711

Convenient check boxes allow you to choose the proper pictogram needed for each chemical in your facility

Designed for a low application temperature

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Description Label
Adhesive Type Permanent Acrylic, Self-adhesive
Color Black/Red/Yellow on White
Finish Gloss
Format Label
Material Vinyl
Printable Sides Single-side Printable
Product Type Label
Languages English
Quantity 50/Pk.
Size (English) 7 x 10 in. (W x H)
Dimensions (L x W) 10 x 7 in.
Width (English) 7 in.
Height (English) 10 in.
Show More Show Less